2,6-Bis[1-(2,6-di-i-propylphenylimino)ethyl]pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Bis-[1-(2,6-diisopropylphenylimino)ethyl]pyridine

A549193-100MG A549193-250MG A549193-50MG
77292-432EA 40 USD
77292-432 77292-434 77292-436
2,6-Bis[1-(2,6-di-i-propylphenylimino)ethyl]pyridine 98%
2,6-Bis[1-(2,6-di-i-propylphenylimino)ethyl]pyridine

Precaución: for Research Use Only

Formula: C₃₃H₄₃N₃
MDL Number: MFCD01862436
CAS Number: 204203-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR