tert-Butyl 2-phenylpiperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

A548965-1G A548965-250MG A548965-5G A548965-100MG
77279-972EA 239 USD
77279-972 77279-974 77279-976 77749-140
tert-Butyl 2-phenylpiperazine-1-carboxylate 98%
tert-Butyl 2-phenylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₂O₂
MDL Number: MFCD07367734
CAS Number: 886766-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR