Dipropyl malonate 98%

Supplier: Ambeed
Warning

A548771-100G A548771-1G A548771-250MG A548771-25G A548771-5G
77266-198EA 406.85 USD
77266-198 77266-200 77266-202 77266-204 77266-206
Dipropyl malonate 98%
Dipropyl malonate

Precaución: for Research Use Only

Formula: C₉H₁₆O₄
MDL Number: MFCD00059406
CAS Number: 1117-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR