1-(2,4-Dimethoxyphenyl)methanamine hydrochloride 98%

Supplier: Ambeed
Warning

A548729-100G A548729-25G A548729-5G
77253-790EA 183 USD
77253-790 77253-792 77253-794
1-(2,4-Dimethoxyphenyl)methanamine hydrochloride 98%
1-(2,4-Dimethoxyphenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄ClNO₂
MDL Number: MFCD00012854
CAS Number: 20781-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR