3,4-Dibromothiophene-2,5-dicarboxaldehyde 95+%

Supplier: Ambeed
Danger

Synonyms: 3,4-Dibromo-2,5-dicarboxaldehyde

A548035-100MG A548035-1G A548035-250MG
77279-966EA 57 USD
77279-966 77279-968 77279-970
3,4-Dibromothiophene-2,5-dicarboxaldehyde 95+%
3,4-Dibromothiophene-2,5-dicarboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₂Br₂O₂S
MDL Number: MFCD11656162
CAS Number: 25373-20-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR