6-Isopropylpyrimidine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A547997-100MG A547997-250MG
77253-784EA 19.57 USD
77253-784 77253-786
6-Isopropylpyrimidine-2,4(1H,3H)-dione 97%
6-Isopropylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.17 g/mol
MDL Number: MFCD21234689
CAS Number: 58289-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR