Ethyl 4-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97%

Supplier: Ambeed
Warning

A547363-100MG A547363-1G A547363-250MG
77266-176EA 35.02 USD
77266-176 77266-178 77266-180
Ethyl 4-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97%
Ethyl 4-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₅H₂₀BClO₄
MW: 310.58 g/mol
MDL Number: MFCD22494062
CAS Number: 1146214-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR