2-Methoxyethyl methacrylate 98% stabilized

Supplier: Ambeed
Warning

Synonyms: Ethylene Glycol Monomethyl Ether Methacrylate, Methacrylic Acid 2-Methoxyethyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A547163-1000G A547163-100G A547163-25G A547163-500G
77266-168EA 78.28 USD
77266-168 77266-170 77266-172 77266-174
2-Methoxyethyl methacrylate 98% stabilized
2-Methoxyethyl methacrylate

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD00048122
CAS Number: 6976-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR