3,4-Difluorophenol 98%

Supplier: Ambeed
Danger

A547085-10G A547085-25G A547085-5G
77292-426EA 11.33 USD
77292-426 77292-428 77572-738
3,4-Difluorophenol 98%
3,4-Difluorophenol

Precaución: for Research Use Only

Formula: C₆H₄F₂O
MW: 130.09 g/mol
MDL Number: MFCD00010315
CAS Number: 2713-33-9
UN: 2921
ADR: 8,4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR