3-(Trifluoromethyl)-p-anisidine 98%

Supplier: Ambeed
Danger

Synonyms: 3-(Trifluoromethyl)-4-anisidine, 5-Amino-2-methoxybenzotrifluoride, 4-Methoxy-3-(trifluoromethyl)aniline, 3-(Trifluoromethyl)-4-methoxyaniline, 4-Amino-2-(trifluoromethyl)anisole

A546575-10G A546575-1G A546575-5G A546575-25G
77253-774EA 30.9 USD
77253-774 77253-776 77253-778 77572-736
3-(Trifluoromethyl)-p-anisidine 98%
3-(Trifluoromethyl)-p-anisidine

Precaución: for Research Use Only

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
MDL Number: MFCD00043460
CAS Number: 393-15-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR