2-(4-Isopropoxyphenyl)acetic acid 97%

Supplier: Ambeed
Warning

A545302-1G A545302-250MG A545302-5G
77253-764EA 21.99 USD
77253-764 77253-766 77253-768
2-(4-Isopropoxyphenyl)acetic acid 97%
2-(4-Isopropoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MDL Number: MFCD01050396
CAS Number: 55784-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR