(S)-2,2'-Bis[bis(3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (S)-Xylyl-Garphos, (S)-2,2'-Bis[bis(3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxy)-1,1'-biphenyl, (S)-(4,4',6,6'-Tetramethoxybiphenyl-2,2'-diyl)bis[bis(3,5-dimethylphenyl)phosphine]

A545247-100MG A545247-1G A545247-250MG A545247-50MG
77253-756EA 73 USD
77253-756 77253-758 77253-760 77253-762
(S)-2,2'-Bis[bis(3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl ≥98%, ee 99%
(S)-2,2'-Bis[bis(3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl

Precaución: for Research Use Only

Formula: C₄₈H₅₂O₄P₂
MDL Number: MFCD19443624
CAS Number: 1365531-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR