1H-Pyrrolo[2,3-b]pyridine-3-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 7-Azaindole-3-carbonitrile, 3-Cyano-1H-pyrrolo[2,3-b]pyridine,3-Cyano-7-azaindole

A545219-100MG A545219-10G A545219-1G A545219-250MG A545219-25G A545219-5G
77241-924EA 5.15 USD
77241-924 77241-926 77241-928 77241-930 77241-932 77241-934
1H-Pyrrolo[2,3-b]pyridine-3-carbonitrile 97%
1H-Pyrrolo[2,3-b]pyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₅N₃
MW: 143.15 g/mol
MDL Number: MFCD07778359
CAS Number: 4414-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR