[1,1':4',1'':4'',1'''-Quaterphenyl]-4,4'''-diamine 98%

Supplier: Ambeed
Danger

A544908-100MG A544908-5G A544908-1G
77253-752EA 25.75 USD
77253-752 77253-754 77617-324
[1,1':4',1'':4'',1'''-Quaterphenyl]-4,4'''-diamine 98%
[1,1':4',1'':4'',1'''-Quaterphenyl]-4,4'''-diamine

Precaución: for Research Use Only

Formula: C₂₄H₂₀N₂
MW: 336.42 g/mol
MDL Number: MFCD02752104
CAS Number: 53693-67-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR