5-Bromo-4-chloro-2-fluoroaniline 95%

Supplier: Ambeed
Warning

A544560-1G A544560-250MG A544560-5G A544560-25G
77253-748EA 24 USD
77253-748 77253-750 77551-648 77747-111
5-Bromo-4-chloro-2-fluoroaniline 95%
5-Bromo-4-chloro-2-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrClFN
MDL Number: MFCD00274773
CAS Number: 111010-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR