4-(4-Chlorophenoxy)butyric acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Chlorophenoxy)butanoic acid

A544456-1G A544456-25G A544456-5G
77292-410EA 10.3 USD
77292-410 77292-412 77292-414
4-(4-Chlorophenoxy)butyric acid 95%
4-(4-Chlorophenoxy)butyric acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
MW: 214.65 g/mol
MDL Number: MFCD00040911
CAS Number: 3547-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR