2-(Oxazolidin-3-yl)ethanol 95%

Supplier: Ambeed
Warning

A544357-100MG A544357-1G A544357-250MG
77241-906EA 21 USD
77241-906 77241-908 77241-910
2-(Oxazolidin-3-yl)ethanol 95%
2-(Oxazolidin-3-yl)ethanol

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MDL Number: MFCD00043560
CAS Number: 20073-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR