2,5-Bis(trifluoromethyl)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Bis(trifluoromethyl)phenylboronic acid

A544285-1G A544285-250MG A544285-5G
77253-740EA 78.28 USD
77253-740 77253-742 77253-744
2,5-Bis(trifluoromethyl)benzeneboronic acid 98%
2,5-Bis(trifluoromethyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₈H₅BF₆O₂
MW: 257.926 g/mol
MDL Number: MFCD04038223
CAS Number: 196083-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR