(2R,2'R)-(+)-[N,N'-Bis(2-pyridylmethyl)]-2,2'-bipyrrolidine tetrahydrochloride 98%

Supplier: Ambeed
Warning

A544141-100MG A544141-1G A544141-250MG
77253-734EA 167.89 USD
77253-734 77253-736 77253-738
(2R,2'R)-(+)-[N,N'-Bis(2-pyridylmethyl)]-2,2'-bipyrrolidine tetrahydrochloride 98%
(2R,2'R)-(+)-[N,N'-Bis(2-pyridylmethyl)]-2,2'-bipyrrolidine tetrahydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₃₀Cl₄N₄
MW: 468.29 g/mol
MDL Number: MFCD16621416
CAS Number: 1228077-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR