6-Chloroindole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A544131-1G A544131-250MG A544131-25G A544131-5G
77279-954EA 23 USD
77279-954 77279-956 77279-958 77617-318
6-Chloroindole-2-carboxylic acid 98%
6-Chloroindole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂
MDL Number: MFCD01863157
CAS Number: 16732-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR