(4S,4'S)-2,2'-(4-Bromopyridine-2,6-diyl)bis(4-isopropyl-4,5-dihydrooxazole) 98%

Supplier: Ambeed
Warning

A543853-100MG A543853-250MG
77241-896EA 145.23 USD
77241-896 77241-898
(4S,4'S)-2,2'-(4-Bromopyridine-2,6-diyl)bis(4-isopropyl-4,5-dihydrooxazole) 98%
(4S,4'S)-2,2'-(4-Bromopyridine-2,6-diyl)bis(4-isopropyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₁₇H₂₂BrN₃O₂
MW: 380.28 g/mol
CAS Number: 477351-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR