3-Chloro-1-iodopropane 98% stabilized

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-3-iodopropane

Stabilization: Stabilized with Copper chip

A543592-100G A543592-10G A543592-25G A543592-500G A543592-5G
77241-886EA 109.18 USD
77241-886 77241-888 77241-890 77241-892 77241-894
3-Chloro-1-iodopropane 98% stabilized
3-Chloro-1-iodopropane

Precaución: for Research Use Only

Formula: C₃H₆ClI
MW: 204.44 g/mol
MDL Number: MFCD00001096
CAS Number: 6940-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR