2,3,6,7,10,11-Hexabromotriphenylene 98%

Supplier: Ambeed
Warning

A543371-100MG A543371-10G A543371-1G A543371-250MG A543371-5G
77253-724EA 33.99 USD
77253-724 77253-726 77253-728 77253-730 77253-732
2,3,6,7,10,11-Hexabromotriphenylene 98%
2,3,6,7,10,11-Hexabromotriphenylene

Precaución: for Research Use Only

Formula: C₁₈H₆Br₆
MW: 701.67 g/mol
MDL Number: MFCD09834716
CAS Number: 82632-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR