(E)-N-(3-((5-Chloro-4-(1H-indol-3-yl)pyrimidin-2-yl)amino)phenyl)-3-(4-(dimethylamino)but-2-enamido)benzamide ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
THZ2
A543298-10MG
A543298-1MG
A543298-5MG
77241-880EA
63.86
USD
77241-880
77241-882
77241-884
(E)-N-(3-((5-Chloro-4-(1H-indol-3-yl)pyrimidin-2-yl)amino)phenyl)-3-(4-(dimethylamino)but-2-enamido)benzamide ≥98%
(E)-N-(3-((5-Chloro-4-(1H-indol-3-yl)pyrimidin-2-yl)amino)phenyl)-3-(4-(dimethylamino)but-2-enamido)benzamide
Precaución: for Research Use Only
Formula:
C₃₁H₂₈ClN₇O₂ MW: 566.05 g/mol |
MDL Number:
MFCD28716099 CAS Number: 1604810-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...