(3-Methoxy-4-((tetrahydro-2H-pyran-4-yl)oxy)phenyl)methanamine 97%

Supplier: Ambeed
Warning

A543106-1G A543106-250MG
77241-876EA 367.71 USD
77241-876 77241-878
(3-Methoxy-4-((tetrahydro-2H-pyran-4-yl)oxy)phenyl)methanamine 97%
(3-Methoxy-4-((tetrahydro-2H-pyran-4-yl)oxy)phenyl)methanamine

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₃
MW: 237.29 g/mol
MDL Number: MFCD19059753
CAS Number: 1378892-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR