2-Bromo-2'-(trifluoromethyl)acetophenone 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-[2-(trifluoromethyl)phenyl]-1-ethanone

A542491-10G A542491-1G A542491-250MG A542491-5G
77253-706EA 353 USD
77253-706 77253-708 77253-710 77253-712
2-Bromo-2'-(trifluoromethyl)acetophenone 98%
2-Bromo-2'-(trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O
MDL Number: MFCD03094304
CAS Number: 54109-16-9
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR