tert-Butyl ((S)-1-((2S,4R)-4-((7-chloro-4-methoxyisoquinolin-1-yl)oxy)-2-(((1R,2S)-1-((cyclopropylsulfonyl)carbamoyl)-2-vinylcyclopropyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: Asunaprevir

A542126-10MG A542126-50MG A542126-5MG
77241-870EA 65.92 USD
77241-870 77241-872 77241-874
tert-Butyl ((S)-1-((2S,4R)-4-((7-chloro-4-methoxyisoquinolin-1-yl)oxy)-2-(((1R,2S)-1-((cyclopropylsulfonyl)carbamoyl)-2-vinylcyclopropyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)carbamate 98%
tert-Butyl ((S)-1-((2S,4R)-4-((7-chloro-4-methoxyisoquinolin-1-yl)oxy)-2-(((1R,2S)-1-((cyclopropylsulphonyl)carbamoyl)-2-vinylcyclopropyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₃₅H₄₆ClN₅O₉S
MW: 748.29 g/mol
MDL Number: MFCD27987900
CAS Number: 630420-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR