2-(4-Methoxyphenyl)-2-phenylacetonitrile 97%

Supplier: Ambeed
Warning

A542074-100MG A542074-1G A542074-250MG
77241-864EA 25.75 USD
77241-864 77241-866 77241-868
2-(4-Methoxyphenyl)-2-phenylacetonitrile 97%
2-(4-Methoxyphenyl)-2-phenylacetonitrile

Precaución: for Research Use Only

Formula: C₁₅H₁₃NO
MW: 223.26 g/mol
MDL Number: MFCD19302710
CAS Number: 4578-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR