3-Benzyl-4-methylthiazolium chloride 95+%

Supplier: Ambeed
Warning

Synonyms: 3-Benzyl-4-methylthiazol-3-ium chloride

A541494-1G A541494-250MG A541494-5G
77279-932EA 24 USD
77279-932 77279-934 77279-936
3-Benzyl-4-methylthiazolium chloride 95+%
3-Benzyl-4-methylthiazolium chloride

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClNS
MDL Number: MFCD01318305
CAS Number: 4209-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR