2-Amino-4,5-dimethoxybenzonitrile 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Cyano-4,5-dimethoxyaniline
A541445-100G
A541445-25G
A541445-5G
A541445-1G
77253-694EA
719.97
USD
77253-694
77253-696
77253-698
77747-108
2-Amino-4,5-dimethoxybenzonitrile 98%
2-Amino-4,5-dimethoxybenzonitrile
Precaución: for Research Use Only
Formula:
C₉H₁₀N₂O₂ MW: 178.19 g/mol |
MDL Number:
MFCD00058682 CAS Number: 26961-27-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...