2-Chloroethyl methyl ether 98%

Supplier: Ambeed
Danger

Synonyms: 2-Methoxyethyl chloride

A541311-10G A541311-25G A541311-5G
77266-128EA 8.24 USD
77266-128 77266-130 77266-132
2-Chloroethyl methyl ether 98%
2-Chloroethyl methyl ether

Precaución: for Research Use Only

Formula: C₃H₇ClO
MW: 94.54 g/mol
MDL Number: MFCD00000972
CAS Number: 627-42-9
UN: 3271
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR