(2S,3S,4R,5S,6R)-3-Acetamido-6-(acetoxymethyl)tetrahydro-2H-pyran-2,4,5-triyl triacetate 97%
Supplier: Ambeed
|
Warning
|
A541217-100MG
A541217-1G
A541217-250MG
77266-122EA
29
USD
77266-122
77266-124
77266-126
(2S,3S,4R,5S,6R)-3-Acetamido-6-(acetoxymethyl)tetrahydro-2H-pyran-2,4,5-triyl triacetate 97%
(2S,3S,4R,5S,6R)-3-Acetamido-6-(acetoxymethyl)tetrahydro-2H-pyran-2,4,5-triyl triacetate
Precaución: for Research Use Only
Formula:
C₁₆H₂₃NO₁₀ |
MDL Number:
MFCD30491713 CAS Number: 6730-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...