2,3,5,6-Tetrafluoro-4-methoxybenzenesulfonylchloride 95%

Supplier: Ambeed
Danger

A540569-100MG A540569-1G A540569-250MG
77253-686EA 50 USD
77253-686 77253-688 77253-690
2,3,5,6-Tetrafluoro-4-methoxybenzenesulfonylchloride 95%
2,3,5,6-Tetrafluoro-4-methoxybenzenesulfonylchloride

Precaución: for Research Use Only

Formula: C₇H₃ClF₄O₃S
MDL Number: MFCD06660766
CAS Number: 40586-69-4
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR