Bis(triphenylphosphino)(2-methylphenyl)chloronickel(II) 98%

Supplier: Ambeed
Danger

A540524-100MG A540524-1G A540524-250MG
77253-680EA 20 USD
77253-680 77253-682 77253-684
Bis(triphenylphosphino)(2-methylphenyl)chloronickel(II) 98%
Bis(triphenylphosphino)(2-methylphenyl)chloronickel(II)

Precaución: for Research Use Only

Formula: C₄₃H₃₇ClNiP₂
MW: 709.84 g/mol
MDL Number: MFCD28144558
CAS Number: 27057-09-6
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR