Leuco crystal violet 98%

Supplier: Ambeed
Warning

Synonyms: 4,4',4''-Methanetriyltris(N,N-dimethylaniline), Tris(4-dimethylaminophenyl)methane, 4,4',4''-Methylidynetris(N,N-dimethylaniline)

A540182-100G A540182-25G
77292-380EA 17 USD
77292-380 77292-382 77292-384
Leuco crystal violet 98%
Leuco crystal violet

Precaución: for Research Use Only

Formula: C₂₅H₃₁N₃
MW: 373.54 g/mol
MDL Number: MFCD00008314
CAS Number: 603-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR