(+)-trans-1,2-Cyclohexanedicarboxylic anhydride 98%

Supplier: Ambeed
Danger

Synonyms: (3aR,7aR)-Hexahydroisobenzofuran-1,3-dione

A539735-100MG A539735-250MG
77266-106EA 74.16 USD
77266-106 77266-108
(+)-trans-1,2-Cyclohexanedicarboxylic anhydride 98%
(+)-trans-1,2-Cyclohexanedicarboxylic anhydride

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
MW: 154.16 g/mol
MDL Number: MFCD00674194
CAS Number: 71749-03-6
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR