NQ301 98%

Supplier: Ambeed
Danger

Synonyms: 2-((4-Acetylphenyl)amino)-3-chloronaphthalene-1,4-dione

A539547-100MG A539547-10MG A539547-1MG A539547-25MG A539547-50MG A539547-5MG
77266-094EA 130 USD
77266-094 77266-096 77266-098 77266-100 77266-102 77266-104
NQ301 98%
NQ301

Precaución: for Research Use Only

Formula: C₁₈H₁₂ClNO₃
MDL Number: MFCD00559986
CAS Number: 130089-98-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR