4-Bromomethyl-2'-fluorobiphenyl 97%

Supplier: Ambeed
Danger

Synonyms: 2-fluoro-4'-bromomethylbiphenyl, 4'-(Bromomethyl)-2-fluoro-1,1'-biphenyl

A539537-1G A539537-250MG
77241-856EA 293.55 USD
77241-856 77241-858
4-Bromomethyl-2'-fluorobiphenyl 97%
4-Bromomethyl-2'-fluorobiphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrF
MW: 265.13 g/mol
MDL Number: MFCD15146811
CAS Number: 193013-76-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR