2,6-Dimethoxybenzoic acid 98%

Supplier: Ambeed
Warning

A539534-100G A539534-25G A539534-5G A539534-500G A539534-100MG
77253-670EA 88.58 USD
77253-670 77253-672 77253-674 77747-104 77747-103
2,6-Dimethoxybenzoic acid 98%
2,6-Dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
MDL Number: MFCD00002437
CAS Number: 1466-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR