4-Isoquinolineboronic acid 98%
Supplier: Ambeed
|
Warning
|
A539363-1G
A539363-25G
A539363-5G
77266-084EA
38.11
USD
77266-084
77266-086
77266-088
4-Isoquinolineboronic acid 98%
4-Isoquinolineboronic acid
Precaución: for Research Use Only
Formula:
C₉H₈BNO₂ MW: 172.9763 g/mol |
MDL Number:
MFCD03412093 CAS Number: 192182-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...