4-Chlorobutanone 97%

Supplier: Ambeed
Danger

Synonyms: 4-Chloro-2-butanone

A539330-1G A539330-5G A539330-10G
77292-376EA 11.33 USD
77292-376 77292-378 77572-716
4-Chlorobutanone 97%
4-Chlorobutanone

Precaución: for Research Use Only

Formula: C₄H₇ClO
MW: 106.55 g/mol
MDL Number: MFCD00037116
CAS Number: 6322-49-2
UN: 1224
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR