α-Bromo-4-chloro-2,6-difluorotoluene 98%

Supplier: Ambeed
Danger

Synonyms: 4-Chloro-2,6-difluorobenzyl bromide, 2-(Bromomethyl)-5-chloro-1,3-difluorobenzene

A539305-10G A539305-1G A539305-250MG
77292-370EA 139.05 USD
77292-370 77292-372 77292-374
α-Bromo-4-chloro-2,6-difluorotoluene 98%
α-Bromo-4-chloro-2,6-difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₄BrClF₂
MW: 241.46 g/mol
MDL Number: MFCD03094463
CAS Number: 537013-52-8
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR