2-(2,2,2-Trimethylacetamido)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Pivaloylamino)phenylboronic acid, 2-(2,2,2-Trimethylacetamido)phenylboronic acid, 2-(tert-Butoxycarbonylamino)phenylboronic acid, 2-(Pivalamido)phenylboronic acid, (2-Pivalamidophenyl)boronic acid

A539137-1G A539137-250MG A539137-5G
77253-660EA 67 USD
77253-660 77253-662 77253-664
2-(2,2,2-Trimethylacetamido)benzeneboronic acid 98%
2-(2,2,2-Trimethylacetamido)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₃
MDL Number: MFCD01114645
CAS Number: 146140-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR