4-Methyl-1-phenyl-2-pentanol 96%

Supplier: Ambeed
Warning

A539045-1G A539045-25G A539045-5G
77292-364EA 8.24 USD
77292-364 77292-366 77292-368
4-Methyl-1-phenyl-2-pentanol 96%
4-Methyl-1-phenyl-2-pentanol

Precaución: for Research Use Only

Formula: C₁₂H₁₈O
MW: 178.28 g/mol
MDL Number: MFCD00209515
CAS Number: 7779-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR