7-Chloro-4-methoxy-1H-indole 98%

Supplier: Ambeed
Warning

A538959-100MG A538959-250MG
77266-080EA 105.06 USD
77266-080 77266-082
7-Chloro-4-methoxy-1H-indole 98%
7-Chloro-4-methoxy-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈ClNO
MW: 181.61 g/mol
MDL Number: MFCD11845601
CAS Number: 948581-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR