4,5-Dimethoxy-2-fluorobenzonitrile 98%

Supplier: Ambeed
Danger

A538342-10G A538342-1G A538342-250MG A538342-5G
77292-356EA 127 USD
77292-356 77292-358 77292-360 77292-362
4,5-Dimethoxy-2-fluorobenzonitrile 98%
4,5-Dimethoxy-2-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₉H₈FNO₂
MDL Number: MFCD00143236
CAS Number: 119396-88-2
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR