N-Phenylglycine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Anilinoacetic acid, 2-(Phenylamino)acetic acid

A538296-1000G A538296-100G A538296-25G A538296-500G A538296-5G
77253-640EA 545.9 USD
77253-640 77253-642 77253-644 77253-646 77253-648
N-Phenylglycine 98%
N-Phenylglycine

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.17 g/mol
MDL Number: MFCD00014009
CAS Number: 103-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR