JNK-IN-7 98+%

Supplier: Ambeed
Warning

Synonyms: 3-[[(E)-4-(dimethylamino)but-2-enoyl]amino]-N-[4-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamide, JNKIN-7, JNK-IN-7, JNK-IN7, JNK IN 7, JNKIN7, 3-(4-(Dimethylamino)but-2-enamido)-N-(4-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)benzamide

A537026-10MG A537026-2MG A537026-1MG A537026-5MG
77241-842EA 44.29 USD
77241-842 77241-844 77617-290 77617-292
JNK-IN-7 98+%
JNK-IN-7

Precaución: for Research Use Only

Formula: C₂₈H₂₇N₇O₂
MW: 493.56 g/mol
MDL Number: MFCD25976765
CAS Number: 1408064-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR