3-Hydroxybicyclo[1.1.1]pentane-1-carboxylic acid 98%

Supplier: Ambeed
Warning

A536506-50MG A536506-250MG A536506-100MG
77266-060EA 214 USD
77266-060 77617-282 77749-120
3-Hydroxybicyclo[1.1.1]pentane-1-carboxylic acid 98%
3-Hydroxybicyclo[1.1.1]pentane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈O₃
MDL Number: MFCD19230741
CAS Number: 83249-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR