4-Bromo-2,6-dichlorobenzenesulfonyl chloride 97%
Supplier: Ambeed
|
Danger
|
A536057-1G
A536057-25G
A536057-5G
77292-344EA
15
USD
77292-344
77292-346
77292-348
4-Bromo-2,6-dichlorobenzenesulfonyl chloride 97%
4-Bromo-2,6-dichlorobenzenesulphonyl chloride
Precaución: for Research Use Only
Formula:
C₆H₂BrCl₃O₂S |
MDL Number:
MFCD03094651 CAS Number: 351003-54-8 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...